AC5424075235 Accela ChemBio Inc.

1-Methyl-3-piperidinone

  • MDL number MFCD07374379
  • Cas Number : 5519-50-6
  • Molecular Weight : 113.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023480&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY023480.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart