AC5424075242 Accela ChemBio Inc.

Benzofuran-2,3-dione

  • MDL number MFCD09035228
  • Cas Number : 4732-72-3
  • Molecular Weight : 148.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023499&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023499.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart