AC5424075301 Accela ChemBio Inc.

Diethyl 4,5-Thiazoledicarboxylate

  • MDL number MFCD01156598
  • Cas Number : 18940-72-2
  • Molecular Weight : 229.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023808&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023808.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart