AC5424075305 Accela ChemBio Inc.

5,6-Dihydro-4H-pyrrolo[3,4-d]thiazole Hydrochloride

  • MDL number MFCD24335251
  • Cas Number : 1434128-58-1
  • Molecular Weight : 162.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023812&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023812.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart