AC5424075310 Accela ChemBio Inc.

2-Ethyl-4-fluoro-1-nitrobenzene

  • MDL number MFCD16658616
  • Cas Number : 1089279-29-7
  • Molecular Weight : 169.1530232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023824&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023824.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart