AC5424075313 Accela ChemBio Inc.

tert-Butyl 2-(1-Methylhydrazinyl)acetate

  • MDL number MFCD20266159
  • Cas Number : 144036-71-5
  • Molecular Weight : 160.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023834&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023834.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart