AC5424075345 Accela ChemBio Inc.

5-Bromo-3-(trifluoroacetyl)indole

  • MDL number MFCD03721051
  • Cas Number : 32387-18-1
  • Molecular Weight : 292.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023971&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023971.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart