AC5424075364 Accela ChemBio Inc.

1-[(2-Fluorophenyl)(4-fluorophenyl)(phenyl)methyl]imidazole

  • MDL number MFCD00865591
  • Cas Number : 119006-77-8
  • Molecular Weight : 346.3726464

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024110&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024110.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart