AC5424075368 Accela ChemBio Inc.

3-Fluoro-4-(methylsulfonyl)phenylboronic Acid Pinacol Ester

  • MDL number MFCD18760182
  • Cas Number : 648904-85-2
  • Molecular Weight : 300.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024122&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024122.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart