AC5424075373 Accela ChemBio Inc.

1,4-Dimethyl-5-nitroimidazole

  • MDL number MFCD01721570
  • Cas Number : 57658-79-4
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024132&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024132.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart