AC5424075408 Accela ChemBio Inc.

4-Bromo-2-fluorophenylboronic Acid Pinacol Ester

  • MDL number MFCD22494116
  • Cas Number : 1326316-85-1
  • Molecular Weight : 300.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024422&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024422.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart