AC5424075415 Accela ChemBio Inc.

1,3-Bis[4-(tert-butyl)phenyl]-2,2-Bis[(S)-4-phenyl-4,5-dihydro-2-oxazolyl]propane

  • MDL number MFCD26404519
  • Cas Number : 1428328-51-1
  • Molecular Weight : 598.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024444&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024444.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart