AC5424075416 Accela ChemBio Inc.

2-Amino-2',5'-dimethoxyacetophenone Hydrochloride

  • MDL number MFCD12911640
  • Cas Number : 671224-08-1
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024453&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024453.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart