AC5424075425 Accela ChemBio Inc.

Ethyl 1-Ethylpyrazole-4-carboxylate

  • MDL number MFCD16620009
  • Cas Number : 447401-91-4
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024516&attname=GoodsCode

Safety Information

hcodes No GHS SYMBOl
MSDS http://pdf.accelachem.com/cn/msds/SY024516.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart