AC5424075453 Accela ChemBio Inc.

3-(Hydroxymethyl)imidazo[1,2-a]pyridine

  • MDL number MFCD09864457
  • Cas Number : 30489-43-1
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024778&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024778.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart