AC5424075462 Accela ChemBio Inc.

2-(Hydroxymethyl)quinoxaline

  • MDL number MFCD00781872
  • Cas Number : 41242-94-8
  • Molecular Weight : 160.17262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024806&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024806.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart