AC5424075481 Accela ChemBio Inc.

1H-Indene-2-carboxylic Acid

  • MDL number MFCD00039586
  • Cas Number : 41712-14-5
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024866&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024866.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart