AC5424075555 Accela ChemBio Inc.

4-(tert-Butoxy)benzamide

  • MDL number MFCD03840534
  • Cas Number : 99985-67-8
  • Molecular Weight : 193.2423

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025202&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025202.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart