AC5424075564 Accela ChemBio Inc.

Ethyl 2-(4,6-Dihydroxy-5-pyrimidyl)acetate

  • MDL number MFCD20527459
  • Cas Number : 1095822-20-0
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025213&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025213.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart