AC5424075565 Accela ChemBio Inc.

Ethyl 2-(4,6-Dichloro-5-pyrimidyl)acetate

  • MDL number MFCD13195351
  • Cas Number : 1095822-21-1
  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025214&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025214.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart