AC5424075567 Accela ChemBio Inc.

4-(Dibenzylamino)butanoic Acid

  • MDL number MFCD00889235
  • Cas Number : 106518-44-9
  • Molecular Weight : 283.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025216&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025216.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart