AC5424075569 Accela ChemBio Inc.

4-Amino-2-chloro-5-iodobenzonitrile

  • MDL number MFCD09909319
  • Cas Number : 380241-58-7
  • Molecular Weight : 278.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025218&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025218.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart