AC5424075588 Accela ChemBio Inc.

trans-(2-Aminocyclohexyl)methanol

  • MDL number MFCD09260357
  • Cas Number : 5691-21-4
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025328&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025328.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart