AC5424075606 Accela ChemBio Inc.

4-Methylcyclohexene-1-boronic Acid Pinacol Ester

  • MDL number MFCD11520541
  • Cas Number : 865869-26-7
  • Molecular Weight : 222.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025517&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025517.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart