AC5424075640 Accela ChemBio Inc.

3-Ethylmorpholine-3-carboxylic Acid

  • MDL number MFCD26383905
  • Cas Number : 1519210-56-0
  • Molecular Weight : 159.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025665&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025665.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart