AC5424075655 Accela ChemBio Inc.

1-(Tetrahydropyran-4-yl)piperazine

  • MDL number MFCD11043433
  • Cas Number : 398137-19-4
  • Molecular Weight : 170.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025726&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025726.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart