AC5424075667 Accela ChemBio Inc.

2-(Methylthio)imidazole

  • MDL number MFCD19216815
  • Cas Number : 7666-04-8
  • Molecular Weight : 114.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025774&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025774.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart