AC5424075668 Accela ChemBio Inc.

N-(Cyclopropylmethyl)cyclopropanamine

  • MDL number MFCD11212563
  • Cas Number : 215522-80-8
  • Molecular Weight : 111.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025777&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY025777.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart