AC5424075680 Accela ChemBio Inc.

Dimethyl 3-Aminopyrrole-2,4-dicarboxylate

  • MDL number MFCD10566520
  • Cas Number : 180059-04-5
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025906&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025906.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart