AC5424075688 Accela ChemBio Inc.

4-Formyl-3-hydroxybenzonitrile

  • MDL number MFCD08361770
  • Cas Number : 84102-89-6
  • Molecular Weight : 147.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025920&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025920.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart