AC5424075703 Accela ChemBio Inc.

2-(Allyloxy)benzyl Bromide

  • MDL number MFCD26937549
  • Cas Number : 319918-15-5
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026003&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026003.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart