AC5424075714 Accela ChemBio Inc.

1,4-Bis(Boc)-2-piperazinemethanol

  • MDL number MFCD09908093
  • Cas Number : 143540-05-0
  • Molecular Weight : 316.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026031&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026031.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart