AC5424075729 Accela ChemBio Inc.

(1S,2S)-2-Aminocyclohexanol

  • MDL number MFCD08061326
  • Cas Number : 74111-21-0
  • Molecular Weight : 115.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026110&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY026110.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart