AC5424075735 Accela ChemBio Inc.

4-(Aminomethyl)-2,2-dimethyl-1,3-benzodioxole

  • MDL number MFCD27500636
  • Cas Number : 1823947-30-3
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026120&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026120.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart