AC5424075742 Accela ChemBio Inc.

1-[2-[(Tetrahydropyran-4-yl)oxy]phenyl]methylamine

  • MDL number MFCD09025871
  • Cas Number : 898289-33-3
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026152&attname=GoodsCode

Safety Information

signalword Danger
pcodes P280-P310-P301 + P330 + P331-P305 + P351 + P338
hcodes GHS05
MSDS http://pdf.accelachem.com/cn/msds/SY026152.pdf
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart