AC5424075754 Accela ChemBio Inc.

3,5-Dibromo-4-chlorophenol

  • MDL number MFCD18914596
  • Cas Number : 74104-19-1
  • Molecular Weight : 286.34842

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026185&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026185.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart