AC5424075758 Accela ChemBio Inc.

3-Fluoro-5-(methoxycarbonyl)benzoic Acid

  • MDL number MFCD08458981
  • Cas Number : 660416-36-4
  • Molecular Weight : 198.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026333&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026333.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart