AC5424075763 Accela ChemBio Inc.

Cbz-D-allo-isoleucine

  • MDL number MFCD02094394
  • Cas Number : 55723-45-0
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026339&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026339.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart