AC5424075764 Accela ChemBio Inc.

D-allo-Isoleucine Ethyl Ester Hydrochloride

  • MDL number MFCD26523292
  • Cas Number : 315700-65-3
  • Molecular Weight : 195.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026343&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026343.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart