AC5424075787 Accela ChemBio Inc.

2-(Methoxymethyl)phenol

  • MDL number MFCD14680991
  • Cas Number : 5635-98-3
  • Molecular Weight : 138.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026429&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026429.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart