AC5424075798 Accela ChemBio Inc.

2-Cyano-3-(4-hydroxyphenyl)propanoic Acid

  • MDL number MFCD11593027
  • Cas Number : 90924-41-7
  • Molecular Weight : 191.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026460&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026460.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart