AC5424075799 Accela ChemBio Inc.

3-(4-Hydroxy-3,5-dimethylphenyl)propanoic Acid

  • MDL number MFCD11188349
  • Cas Number : 7733-58-6
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026461&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026461.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart