AC5424075803 Accela ChemBio Inc.

4-Ethylsalicylaldehyde

  • MDL number MFCD16997668
  • Cas Number : 161876-64-8
  • Molecular Weight : 150.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026477&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026477.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart