AC5424075804 Accela ChemBio Inc.

Methyl 3-(Chloromethyl)-5-nitrobenzoate

  • MDL number MFCD13689045
  • Cas Number : 142320-39-6
  • Molecular Weight : 229.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026478&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026478.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart