AC5424075851 Accela ChemBio Inc.

6-Benzofurancarbaldehyde

  • MDL number MFCD09038098
  • Cas Number : 123297-88-1
  • Molecular Weight : 146.14274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026601&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026601.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart