AC5424075877 Accela ChemBio Inc.

5-Methyl-1,2,4-oxadiazole-3-carboxylic Acid

  • MDL number MFCD08236814
  • Cas Number : 19703-92-5
  • Molecular Weight : 128.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026691&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026691.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart