AC5424075921 Accela ChemBio Inc.

1-(Bromomethyl)-4-methoxycyclohexane

  • MDL number MFCD24459676
  • Cas Number : 141604-51-5
  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026796&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026796.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart