AC5424075931 Accela ChemBio Inc.

2-Amino-5-(chloromethyl)pyrimidine Hydrochloride

  • MDL number MFCD09056752
  • Cas Number : 120747-86-6
  • Molecular Weight : 180.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026827&attname=GoodsCode

Safety Information

signalword Danger
pcodes P201-P281-P308 + P313
MSDS http://pdf.accelachem.com/cn/msds/SY026827.pdf
pictograms GHS08
hstatements H350-H361d
Availability details for
Quantity
  • Availability
  • Add to Cart