AC5424075947 Accela ChemBio Inc.

(E)-3-Bromoacrylic Acid

  • MDL number MFCD00218760
  • Cas Number : 6213-89-4
  • Molecular Weight : 150.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026854&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026854.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart