AC5424075958 Accela ChemBio Inc.

2-(Aminomethyl)-1,3-benzodioxole

  • MDL number MFCD18805479
  • Cas Number : 936-78-7
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026998&attname=GoodsCode

Safety Information

hcodes No GHS SYMBOl
MSDS http://pdf.accelachem.com/cn/msds/SY026998.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart